3-Brom-N-propyl-5-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBHC-6453-1GEA 135 EUR
COBBHC-6453-1G COBBHC-6453-5G
3-Brom-N-propyl-5-methylbenzolsulfonamid
3-Brom-N-propyl-5-methylbenzolsulfonamid
Formel: C₁₀H₁₄BrNO₂S
MDL: MFCD09972145
CAS-Nummer: 1020252-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR