3-Brom-N-tert-butyl-4-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS INC.
COBBOT-0164-1GEA
118
EUR
COBBOT-0164-1G
COBBOT-0164-5G
3-Brom-N-tert-butyl-4-methylbenzolsulfonamid
3-Brom-N-tert-butyl-4-methylbenzolsulfonamid
Formel:
C₁₁H₁₆BrNO₂S |
MDL:
MFCD07363835 CAS-Nummer: 850429-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...