Sulfamethoxypyridazin

Lieferant: EDQM

EDQMS2150000EA 195 EUR
EDQMS2150000
Sulfamethoxypyridazin
Sulfamethoxypyridazin
Formel: C₁₁H₁₂N₄O₃S
MDL: MFCD00057372
CAS-Nummer: 80-35-3
EINECS: 201-272-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR