N-Cyclohexyl-4-fluorbenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBOT-0961-5GEA 143 EUR
COBBOT-0961-5G
N-Cyclohexyl-4-fluorbenzolsulfonamid
N-Cyclohexyl-4-fluorbenzolsulfonamid
Formel: C₁₂H₁₆FNO₂S
MDL: MFCD01213082
CAS-Nummer: 565-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR