N-Ethyl-3-fluorbenzamid
Lieferant: COMBI-BLOCKS INC.
COBBCA-4766-5GEA
143
EUR
COBBCA-4766-5G
N-Ethyl-3-fluorbenzamid
N-Ethyl-3-fluorbenzamid
Formel:
C₉H₁₀FNO |
MDL:
MFCD01338769 CAS-Nummer: 150079-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...