3-Fluor-N-isopropylbenzamid

Lieferant: COMBI-BLOCKS INC.

COBBCA-4767-5GEA 143 EUR
COBBCA-4767-5G
3-Fluor-N-isopropylbenzamid
3-Fluor-N-isopropylbenzamid
Formel: C₁₀H₁₂FNO
MDL: MFCD01341858
CAS-Nummer: 459218-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR