3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure ≥98%
|
Achtung
|
H52596.03EA
291
EUR
H52596.03
H52596.MD
3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure ≥98%
3-Fluor-4-(2-hydroxyethylcarbamoyl)phenylboronsäure
Formel:
C₉H₁₁BFNO₄ Molecular Weight: 227,00 g/mol Schmelzpunkt: 217…219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689535 CAS-Nummer: 874289-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...