3-Amino-N,N-diethylbenzamid

Lieferant: COMBI-BLOCKS INC.

COBBST-9632-1GEA 164 EUR
COBBST-9632-1G
3-Amino-N,N-diethylbenzamid
3-Amino-N,N-diethylbenzamid
Formel: C₁₁H₁₆N₂O
MDL: MFCD00025279
CAS-Nummer: 68269-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR