4-Chlor-2-methoxy-N-methylanilin

Lieferant: COMBI-BLOCKS INC.

COBBAN-1170-1GEA 713 EUR
COBBAN-1170-1G
4-Chlor-2-methoxy-N-methylanilin
4-Chlor-2-methoxy-N-methylanilin
Formel: C₈H₁₀ClNO
MDL: MFCD00027506
CAS-Nummer: 35122-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR