5-Chlor-2-(trifluormethoxy)benzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

H31584.06EA 379 EUR
H31584.06 H31584.03
5-Chlor-2-(trifluormethoxy)benzaldehyd ≥97%
5-Chlor-2-(trifluormethoxy)benzaldehyd
Formel: C₈H₄ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11519353
CAS-Nummer: 1092461-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR