3-Chlor-4-(methylcarbamoyl)phenylboronsäure 96%
Lieferant: Thermo Scientific
|
Achtung
|
H52721.MDEA
125
EUR
H52721.MD
3-Chlor-4-(methylcarbamoyl)phenylboronsäure 96%
3-Chlor-4-(methylcarbamoyl)phenylboronsäure
Formel:
C₈H₉BClNO₃ Molecular Weight: 213,43 g/mol Schmelzpunkt: 164…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363766 CAS-Nummer: 850589-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...