2-Brom-N-cyclohexylbenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBHC-6153-5GEA 255 EUR
COBBHC-6153-5G COBBHC-6153-1G
2-Brom-N-cyclohexylbenzolsulfonamid
2-Brom-N-cyclohexylbenzolsulfonamid
Formel: C₁₂H₁₆BrNO₂S
MDL: MFCD09800965
CAS-Nummer: 951883-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR