4-Brom-N-(2-hydroxyethyl)benzolsulfonamid
Lieferant: COMBI-BLOCKS INC.
COBBOT-0085-5GEA
601
EUR
COBBOT-0085-5G
COBBOT-0085-1G
4-Brom-N-(2-hydroxyethyl)benzolsulfonamid
4-Brom-N-(2-hydroxyethyl)benzolsulfonamid
Formel:
C₈H₁₀BrNO₃S |
MDL:
MFCD03470703 CAS-Nummer: 59724-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...