2-Brom-N,N-diisopropyl-5-fluorbenzamid

Lieferant: COMBI-BLOCKS INC.

COBBCA-4442-5GEA 432 EUR
COBBCA-4442-5G COBBCA-4442-1G
2-Brom-N,N-diisopropyl-5-fluorbenzamid
2-Brom-N,N-diisopropyl-5-fluorbenzamid
Formel: C₁₃H₁₇BrFNO
MDL: MFCD09800912
CAS-Nummer: 951884-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR