3-Brom-5-nitro-N-propylbenzamid
Lieferant: COMBI-BLOCKS INC.
COBBCA-4359-5GEA
291
EUR
COBBCA-4359-5G
COBBCA-4359-1G
3-Brom-5-nitro-N-propylbenzamid
3-Brom-5-nitro-N-propylbenzamid
Formel:
C₁₀H₁₁BrN₂O₃ |
MDL:
MFCD09258764 CAS-Nummer: 929000-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...