2-Brom-N-isopropylbenzamid

Lieferant: COMBI-BLOCKS INC.

COBBCS-9409-1GEA 72 EUR
COBBCS-9409-1G COBBCS-9409-5G
2-Brom-N-isopropylbenzamid
2-Brom-N-isopropylbenzamid
Formel: C₁₀H₁₂BrNO
MDL: MFCD00779656
CAS-Nummer: 64141-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR