3-Brom-N,N-diisopropylbenzamid

Lieferant: COMBI-BLOCKS INC.

COBBCA-4484-1GEA 114 EUR
COBBCA-4484-1G COBBCA-4484-5G
3-Brom-N,N-diisopropylbenzamid
3-Brom-N,N-diisopropylbenzamid
Formel: C₁₃H₁₈BrNO
MDL: MFCD00587757
CAS-Nummer: 35309-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR