2-Brom-N,N-dibutylbenzamid
Lieferant: COMBI-BLOCKS INC.
COBBCS-9435-5GEA
143
EUR
COBBCS-9435-5G
COBBCS-9435-1G
2-Brom-N,N-dibutylbenzamid
2-Brom-N,N-dibutylbenzamid
Formel:
C₁₅H₂₂BrNO |
MDL:
MFCD00693779 CAS-Nummer: 349092-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...