5-Brom-2-chlor-N-cyclohexylbenzamid

Lieferant: COMBI-BLOCKS INC.

COBBCA-4722-5GEA 291 EUR
COBBCA-4722-5G COBBCA-4722-1G
5-Brom-2-chlor-N-cyclohexylbenzamid
5-Brom-2-chlor-N-cyclohexylbenzamid
Formel: C₁₃H₁₅BrClNO
MDL: MFCD04220000
CAS-Nummer: 701260-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR