4-Brom-3-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBBB-3231-5GEA
709
EUR
COBBBB-3231-5G
COBBBB-3231-1G
4-Brom-3-nitrophenylboronsäure
4-Brom-3-nitrophenylboronsäure
Formel:
C₆H₅BBrNO₄ Molecular Weight: 245,8200 g/mol |
MDL:
MFCD01074649 CAS-Nummer: 74386-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...