4-Brom-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5834-250MEA 431 EUR
COBBBB-5834-250M
4-Brom-2-nitrophenylboronsäure
4-Brom-2-nitrophenylboronsäure
Formel: C₆H₅BBrNO₄
Molecular Weight: 245,8232 g/mol
MDL: MFCD12025992
CAS-Nummer: 860034-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR