4-(Cyclopropylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5944-250MEA 441 EUR
COBBBB-5944-250M COBBBB-5944-1G
4-(Cyclopropylsulfonyl)phenylboronsäure
4-(Cyclopropylsulfonyl)phenylboronsäure
Formel: C₉H₁₁BO₄S
MDL: MFCD12546569
CAS-Nummer: 1217501-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR