4-Mercaptophenylboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBBB-8372-5GEA
338
EUR
COBBBB-8372-5G
COBBBB-8372-1G
4-Mercaptophenylboronsäure
4-Mercaptophenylboronsäure
Formel:
C₆H₇BO₂S |
MDL:
MFCD03093887 CAS-Nummer: 237429-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...