4-(Methoxycarbamoyl)phenylboronsäure ≥97%
|
Achtung
|
H53188.MDEA
51.6
EUR
H53188.MD
H53188.03
4-(Methoxycarbamoyl)phenylboronsäure ≥97%
4-(Methoxycarbamoyl)phenylboronsäure
Formel:
C₈H₁₀BNO₄ Molecular Weight: 194,98 g/mol Schmelzpunkt: 130…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115670 CAS-Nummer: 850568-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...