3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-3094-1GEA 291 EUR
COBBBB-3094-1G
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure
3-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronsäure
Formel: C₁₀H₁₁BN₂O₃S
Molecular Weight: 250,0800 g/mol
MDL: MFCD07363799
CAS-Nummer: 871333-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR