4-(2-Methoxyethylcarbamoyl)phenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H53155.MDEA
88.2
EUR
H53155.MD
4-(2-Methoxyethylcarbamoyl)phenylboronsäure ≥98%
4-(2-Methoxyethylcarbamoyl)phenylboronsäure
Formel:
C₁₀H₁₄BNO₄ Molecular Weight: 223,04 g/mol Schmelzpunkt: 120…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115682 CAS-Nummer: 850589-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...