4-(Dimethylcarbamoyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

SIAL715492-1GEA 34.8 EUR
SIAL715492-1G
4-(Dimethylcarbamoyl)phenylboronsäure, Sigma-Aldrich®
4-(Dimethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₃
Molecular Weight: 193,01
MDL: MFCD02258943
CAS-Nummer: 405520-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR