2-(2-Chlorphenyl)oxazol
Lieferant: COMBI-BLOCKS INC.
COBBHC-3301-5GEA
432
EUR
COBBHC-3301-5G
COBBHC-3301-1G
2-(2-Chlorphenyl)oxazol
2-(2-Chlorophenyl)oxazole
Formel:
C₉H₆ClNO |
MDL:
MFCD03407330 CAS-Nummer: 62881-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...