2-Methyl-4-thiazolcarbonsäure

Lieferant: Thermo Scientific

MAYBCC05701-0001EA 0 EUR
MAYBCC05701-0001 MAYBCC05701DA MAYBCC05701CB
2-Methyl-4-thiazolcarbonsäure
2-Methyl-4-thiazolecarboxylic acid
Formel: C₅H₅NO₂S
MDL: MFCD03407332
CAS-Nummer: 35272-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR