Ethylthiazol-4-carboxylat, Sigma-Aldrich®

Lieferant: Merck

SIAL707112-500MGEA 27.8 EUR
SIAL707112-500MG
Ethylthiazol-4-carboxylat, Sigma-Aldrich®
Ethylthiazol-4-carboxylat
Formel: C₆H₇NO₂S
Molecular Weight: 157,1900 g/mol
MDL: MFCD03788566
CAS-Nummer: 14527-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR