5-Oxazolcarbonsäure

Lieferant: COMBI-BLOCKS INC.

COBBHI-1271-1GEA 49 EUR
COBBHI-1271-1G
5-Oxazolcarbonsäure
5-Oxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113,07 g/mol
MDL: MFCD04114931
CAS-Nummer: 118994-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR