Ethyl-2-oxazolcarboxylat

Lieferant: CHEMPUR

CHMP56-0022.5EA 0 EUR
CHMP56-0022.5 CHMP56-0022.1
Ethyl-2-oxazolcarboxylat
Ethyl-2-oxazolcarboxylat
Formel: C₆H₇NO₃
MDL: MFCD04114939
CAS-Nummer: 33036-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR