Ethyl-2-chlor-5-oxazolcarboxylat

Lieferant: Apollo Scientific

APOSOR11484-1GEA 109 EUR
APOSOR11484-1G
Ethyl-2-chlor-5-oxazolcarboxylat
Ethyl-2-chlor-5-oxazolcarboxylat
Formel: C₆H₆ClNO₃
Molecular Weight: 175,5697 g/mol
MDL: MFCD09475866
CAS-Nummer: 862599-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR