5-(4-Fluorphenyl)isoxazol ≥99%

H26188.14EA 258 EUR
H26188.14 H26188.06
5-(4-Fluorphenyl)isoxazol ≥99%
5-(4-Fluorophenyl)isoxazole
Formel: C₉H₆FNO
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02183544
CAS-Nummer: 138716-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR