Methyl-5-methylisoxazol-3-carboxylat

Lieferant: Molekula

MOLEM16123593EA 0 EUR
MOLEM16123593
Methyl-5-methylisoxazol-3-carboxylat
Methyl-5-methylisoxazol-3-carboxylat
Formel: C₆H₇NO₃
Molecular Weight: 141,13 g/mol
MDL: MFCD00015895
CAS-Nummer: 19788-35-3
EINECS: 243-311-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR