1H-Indazol-5-carbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL702455-1GEA 41.1 EUR
SIAL702455-1G
1H-Indazol-5-carbonsäure, Sigma-Aldrich®
1H-Indazole-5-carboxylic acid
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
MDL: MFCD06804570
CAS-Nummer: 61700-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR