5-Brom-3-iod-1H-indazol

Lieferant: COMBI-BLOCKS INC.

COBBIN-0271-1GEA 51.8 EUR
COBBIN-0271-1G
5-Brom-3-iod-1H-indazol
5-Bromo-3-iodo-1H-indazole
Formel: C₇H₄BrIN₂
MDL: MFCD07781637
CAS-Nummer: 459133-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR