Zinn(II)tetrafluoroborat 50% in wässriger Lösung

Lieferant: Honeywell Chemicals

HONE01543.9045EA 0 EUR
HONE01543.9045
Zinn(II)tetrafluoroborat 50% in wässriger Lösung
Tin(II) tetrafluoroborate
Formel: B₂F₈Sn
Molecular Weight: 292,30 g/mol
MDL: MFCD00054047
EINECS: 237-487-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR