Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat
Lieferant: Molekula
MOLE43889017-5GEA
0
EUR
MOLE43889017-5G
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat
Formel:
C₇H₁₁N₃O₂ Molecular Weight: 231,26 |
MDL:
MFCD00020731 CAS-Nummer: 16078-71-0 EINECS: 240-224-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...