Ethyl-5-amino-4-cyan-1-(4-methoxyphenyl)-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS INC.

COBBHC-2392-250MEA 0 EUR
COBBHC-2392-250M COBBHC-2392-1G
Ethyl-5-amino-4-cyan-1-(4-methoxyphenyl)-1H-pyrazol-3-carboxylat
Ethyl-5-amino-4-cyan-1-(4-methoxyphenyl)-1H-pyrazol-3-carboxylat
Formel: C₁₄H₁₄N₄O₃
MDL: MFCD09835749
CAS-Nummer: 1150164-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR