Ethyl-5-amino-1-(3-bromphenyl)-4-cyan-1H-pyrazol-3-carboxylat

Lieferant: COMBI-BLOCKS INC.

COBBHC-2295-1GEA 143 EUR
COBBHC-2295-1G
Ethyl-5-amino-1-(3-bromphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Ethyl-5-amino-1-(3-bromphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Formel: C₁₃H₁₁BrN₄O₂
MDL: MFCD09835741
CAS-Nummer: 1150163-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR