Ethyl-5-amino-1-(2-bromphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Lieferant: COMBI-BLOCKS INC.
COBBHC-2296-1GEA
143
EUR
COBBHC-2296-1G
Ethyl-5-amino-1-(2-bromphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Ethyl-5-amino-1-(2-bromphenyl)-4-cyan-1H-pyrazol-3-carboxylat
Formel:
C₁₃H₁₁BrN₄O₂ |
MDL:
MFCD09835742 CAS-Nummer: 1150163-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...