1-Boc-7-(methoxycarbonyl)-2-indolboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5418-1GEA 282 EUR
COBBBB-5418-1G COBBBB-5418-250M
1-Boc-7-(methoxycarbonyl)-2-indolboronsäure
1-Boc-7-(methoxycarbonyl)-2-indolboronsäure
Formel: C₁₅H₁₈BNO₆
MDL: MFCD11504860
CAS-Nummer: 953411-05-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR