5-Methoxy-2-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBFM-2499-250MEA 196 EUR
COBBFM-2499-250M
5-Methoxy-2-indolboronsäurepinakolester
5-Methoxy-2-indolboronsäurepinakolester
Formel: C₁₅H₂₀BNO₃
Molecular Weight: 273,1352 g/mol
MDL: MFCD11858363
CAS-Nummer: 683229-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR