1-Boc-5-cyan-3-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-6215-1GEA 675 EUR
COBBPN-6215-1G
1-Boc-5-cyan-3-indolboronsäurepinakolester
1-Boc-5-cyan-3-indolboronsäurepinakolester
Formel: C₂₀H₂₅BN₂O₄
MDL: MFCD15143604
CAS-Nummer: 1185427-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR