2,6,7-Trichlor-3-(trifluormethyl)chinoxalin

Lieferant: COMBI-BLOCKS INC.

COBBPY-7168-1GEA 830 EUR
COBBPY-7168-1G
2,6,7-Trichlor-3-(trifluormethyl)chinoxalin
2,6,7-Trichloro-3-(trifluoromethyl)quinoxaline
Formel: C₉H₂Cl₃F₃N₂
MDL: MFCD09801052
CAS-Nummer: 281209-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR