2-(N-tert-Butylamino)-3,6,7-trichlorchinoxalin

Lieferant: COMBI-BLOCKS INC.

COBBHC-6208-1GEA 713 EUR
COBBHC-6208-1G
2-(N-tert-Butylamino)-3,6,7-trichlorchinoxalin
2-(N-tert-Butylamino)-3,6,7-trichlorchinoxalin
Formel: C₁₂H₁₂Cl₃N₃
MDL: MFCD09475905
CAS-Nummer: 281211-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR