5-Cyan-1-isopropylbenzimidazol

Lieferant: COMBI-BLOCKS INC.

COBBIN-0209-5GEA 108 EUR
COBBIN-0209-5G
5-Cyan-1-isopropylbenzimidazol
5-Cyan-1-isopropylbenzimidazol
Formel: C₁₁H₁₁N₃
MDL: MFCD15143576
CAS-Nummer: 1215206-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR