4,7-Dichlorthieno[2,3-d]pyridazin

Lieferant: COMBI-BLOCKS INC.

COBBQA-6657-1GEA 431 EUR
COBBQA-6657-1G
4,7-Dichlorthieno[2,3-d]pyridazin
4,7-Dichlorothieno[2,3-d]pyridazine
Formel: C₆H₂Cl₂N₂S
Molecular Weight: 205,0645 g/mol
MDL: MFCD09965522
CAS-Nummer: 699-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR