3-(Trimethylsilylethinyl)pyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL521264-5GEA
76
EUR
SIAL521264-5G
3-(Trimethylsilylethinyl)pyridin, Sigma-Aldrich®
3-(Trimethylsilylethinyl)pyridin
Formel:
C₁₀H₁₃NSi Molecular Weight: 175,31 g/mol |
MDL:
MFCD01863744 CAS-Nummer: 80673-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...